Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Bench top cell sorter, CytoFLEX SRT

CytoFLEX SRT bench top cell sorter, V5-B2-Y5-R0 (12 colors, 3 lasers), 488 nm, 405 nm, 561 nm
Catalog # BECMC71884
Supplier:   Beckman Coulter
Bench top cell sorter, CytoFLEX SRT
Catalog # BECMC71884
Supplier:  Beckman Coulter
Bench top cell sorter, CytoFLEX SRT
Bench top cell sorter, CytoFLEX SRT

CytoFLEX SRT bench top cell sorter, V5-B2-Y5-R0 (12 colors, 3 lasers), 488 nm, 405 nm, 561 nm

Price
Preis anfragen
Anmelden für die Verfügbarkeit
1 * 1 ST
Product Details & Documents

If you need an approachable instrument for sorting multiple populations to generate high quality material for downstream assays, the CytoFLEX SRT benchtop cell sorter is quick to learn and easy to operate. This extension of the CytoFLEX platform is built on the same optical features as the flow cytometer, but equally important, it continues to adhere to the principles that made multicolor applications accessible to many biomedical research, core research, pharma and biotechs.

  • Exquisite sensitivity for multicolor applications, up to 15 fluorescent colors to define populations
  • Parameter matched to the CytoFLEX S V-B-Y-R series
  • Intuitive software to facilitate multicolor analysis, CytExpert for CytoFLEX SRT
  • Uses technology to increase ease of use by automating sort stream setup and maintenance throughout the sort
  • 4-way sorting, mixed mode sorting, ability to catch aborts and preserve precious cells
  • High recovery and post sort viability
  • Accurate single-cell deposition using the Cyclone Movement System, including index sorting
  • Sort into tubes, slides, or microplates

CytoFLEX SRT cell sorter is a benchtop sorter. It is capable of meeting requirements for a wide range of sorting needs. And like the CytoFLEX Platform, it includes innovative technologies that simplify the setup and operation, empowering investigators to focus on the research questions.

The Violet-Blue-Yellow Green-Red (V-B-Y-R) series has 15 fluorescent detectors when fully activated. The fully activated instrument includes five fluorescent channels from the 405 nm (Violet) laser, two from the 488 nm (Blue) laser, five from the 561 nm (Yellow Green) laser, and three from the 638 nm (Red) laser. It can be purchased with as few as five with an option to activate additional lasers and detectors with an activation key.

It is capable of complex sort logic with different combinations of sort settings on each of four streams, including the ability to catch aborts of one of the other streams. The CytoFLEX SRT cell sorter is designed for reliability and supported by teams of experts, remote support is ready using BeckmanConnect for on demand access to our technical specialists, perform many routine maintenance procedures independently.

Additional specifications:
Nozzle diameter: 100 µm
Signal processing: Digital system with 7-decade display
Drop dive frequency: Automatically set, manually adjustable between 30000 - 35000 Hz
Sorting performance: With a target population of 5% and a threshold rate of <10,000 events per second, >99% purity and >80% of Poisson's expected yield
Sorting modes: 4-way sorting; enrichment or purity or single, ability to catch aborts
Collection vessels: 12×75 mm tubes, 15 ml conical tubes, 96-well plates, 96-deep-well plates, 384-well plates, slides
Optical filter: 450/45, 525/40 (2), 585/42, 610/20 (2), 660/10 (2), 675/30, 690/50, 710/50, 712/25, 780/60 (3)

 : CE, RUO. EN12469, BS EN12469, SNS 12469, NF-095, YY-0569, JIS K 3800, AS 1807.1:2009.

 : CytoFLEX SRT is supplied with CyExpert for SRT, computer, monitor and keyboard. An optional Biosafety Cabinet including aerosol evacuation or a standalone Aerosol Evacuation Unit is available.

Spezifikationen:
  • Wavelength:
    488 nm, 405 nm, 561 nm
  • Description:
    CytoFLEX SRT bench top cell sorter, V5-B2-Y5-R0 (12 colors, 3 lasers)
  • Seq:
    07
  • Cat. No.:
    BECMC71884
  • Detection method:
    Avalanche photodiodes
  • Sensitivity:
    <30 MESF for FITC, <10 MESF for PE, <25 MESF for APC
  • Resolution:
    300 nm particles (488 nm SSC), 200 nm particles (405 nm VSSC), rCV 3%
Accessories

Produktinformationen

  • Catalog #
    Availability
    Price
  • Flow-check fluorospheres, 10 μm (nominal diameter) polystyrene fluorescent microspheres suspended in an aqueous medium containing surfactants and preservatives at 1× 106 fluorospheres/ml (nominal concentration)
    Flow-check fluorospheres, 10 μm (nominal diameter) polystyrene fluorescent microspheres suspended in an aqueous medium containing surfactants and preservatives at 1× 106 fluorospheres/ml (nominal concentration)
    BECM6605359
    Anmelden für die Verfügbarkeit
    1 * 3 ST - 373,00 €

Produktinformationen

  • Catalog #
    Availability
    Price
  • Aerosol evacuation system for CytoFLEX SRT, 220/240 V, 50/60 Hz
    Aerosol evacuation system for CytoFLEX SRT, 220/240 V, 50/60 Hz
    BECMC63648
    Anmelden für die Verfügbarkeit
    1 * 1 ST - 12 590,00 €

Produktinformationen

  • Catalog #
    Availability
    Price
  • CytoFLEX SRT nonionic surfactant and azide-free shutdown fluid
    CytoFLEX SRT nonionic surfactant and azide-free shutdown fluid
    BECMC52574
    Anmelden für die Verfügbarkeit
    1 * 10 L - 64,50 €

Produktinformationen

  • Catalog #
    Availability
    Price
  • Contrad 70 cleaning agent, 1×1 liter
    Contrad 70 cleaning agent, 1×1 liter
    BECM81911
    Anmelden für die Verfügbarkeit
    1 * 1 L

Produktinformationen

  • Catalog #
    Availability
    Price
  • IsoFlow sheath fluid
    IsoFlow sheath fluid
    BECM8546859
    Anmelden für die Verfügbarkeit
    1 * 10 L - 42,10 €

Produktinformationen

  • Catalog #
    Availability
    Price
  • CytoFLEX SRT nozzle module, 100 µm
    CytoFLEX SRT nozzle module, 100 µm
    BECMC58834
    Anmelden für die Verfügbarkeit
    1 * 1 ST - 2 110,00 €

Produktinformationen

  • Catalog #
    Availability
    Price
  • CytoFLEX SRT waste air filter
    CytoFLEX SRT waste air filter
    BECMC64210
    Anmelden für die Verfügbarkeit
    1 * 1 ST - 354,00 €

Produktinformationen

  • Catalog #
    Availability
    Price
  • CytoFLEX SRT de-bubble filter
    CytoFLEX SRT de-bubble filter
    BECMC64211
    Anmelden für die Verfügbarkeit
    1 * 1 ST - 114,00 €

Produktinformationen

  • Catalog #
    Availability
    Price
  • CytoFLEX SRT nozzle, 100 µm
    CytoFLEX SRT nozzle, 100 µm
    BECMC68147
    Anmelden für die Verfügbarkeit
    1 * 1 ST - 1 180,00 €

Produktinformationen

  • Catalog #
    Availability
    Price
  • CytoFLEX SRT workstation M720t tower LTSC OS computer
    CytoFLEX SRT workstation M720t tower LTSC OS computer
    BECMC81021
    Anmelden für die Verfügbarkeit
    1 * 1 ST - 6 260,00 €

Produktinformationen

  • Catalog #
    Availability
    Price
  • CytoFLEX SRT 55200 - HP monitor
    CytoFLEX SRT 55200 - HP monitor
    BECMB41724
    Anmelden für die Verfügbarkeit
    1 * 1 ST

Produktinformationen

  • Catalog #
    Availability
    Price
  • CytoFLEX SRT sample line assembly
    CytoFLEX SRT sample line assembly
    BECMC64203
    Anmelden für die Verfügbarkeit
    1 * 1 ST - 672,00 €

Produktinformationen

  • Catalog #
    Availability
    Price
  • CytoFLEX SRT orifice O-ring
    CytoFLEX SRT orifice O-ring
    BECMC64205
    Anmelden für die Verfügbarkeit
    1 * 10 ST - 142,00 €

Produktinformationen

  • Catalog #
    Availability
    Price
  • CytoFLEX SRT capsule filter
    CytoFLEX SRT capsule filter
    BECMC64209
    Anmelden für die Verfügbarkeit
    1 * 1 ST - 354,00 €