ContactUsLinkComponent
Human CD3E&CD3G Heterodimer Protein, Fc Tag&Fc Tag (CDG-H5253) is expressed from human 293 cells (HEK293). It contains AA Asp 23 - A sp 126 (CD3E) & Gln 23 - Ser 116 (CD3G) (Accession # NP_000724.1 (CD3E) & AAI13831 (CD3G)). 1 * 100 µG
Catalog # ACRBCDG-H5253100UG
Supplier:  ACROBIOSYSTEMS
Human CD3E&CD3G Heterodimer Protein, Fc Tag&Fc Tag (CDG-H5253) is expressed from human 293 cells (HEK293). It contains AA Asp 23 - A sp 126 (CD3E) & Gln 23 - Ser 116 (CD3G) (Accession # NP_000724.1 (CD3E) & AAI13831 (CD3G)). 1 * 100 µG
Catalog # ACRBCDG-H5253100UG
Supplier:  ACROBIOSYSTEMS