ContactUsLinkComponent
HCoV-HKU1 (isolate N5) Spike Trimer, His Tag (SPN-H52H5) is the ectodomain of HCoV-HKU1 (isolate N5) spike protein which contains AA Ala 13 - Asn 1276 (Accession # Q0ZME7-1). The recombinant protein is expressed from human 293 cells (HEK293) with T4 1 * 50 µG
Catalog # ACRBSPN-H52H5-50UG
Supplier:  ACROBIOSYSTEMS
HCoV-HKU1 (isolate N5) Spike Trimer, His Tag (SPN-H52H5) is the ectodomain of HCoV-HKU1 (isolate N5) spike protein which contains AA Ala 13 - Asn 1276 (Accession # Q0ZME7-1). The recombinant protein is expressed from human 293 cells (HEK293) with T4 1 * 50 µG
Catalog # ACRBSPN-H52H5-50UG
Supplier:  ACROBIOSYSTEMS