ContactUsLinkComponent
Mouse MCAM, His Tag (MCM-M52H3) is expressed from human 293 cells (HEK293). It contains AA Val 24 - Val 563 (Accession # Q8R2Y2-1). 1 * 100 µG
Catalog # ACRBMCM-M52H3100UG
Lieferant:  ACRO Biosystems
Mouse MCAM, His Tag (MCM-M52H3) is expressed from human 293 cells (HEK293). It contains AA Val 24 - Val 563 (Accession # Q8R2Y2-1). 1 * 100 µG
Catalog # ACRBMCM-M52H3100UG
Lieferant:  ACRO Biosystems
Frequently Bought Together