ContactUsLinkComponent
Human Galectin-1, Tag Free (GA1-H4113) is expressed from E.coli cells. It contains AA Met 1 - Asp 135 (Accession # P09382-1). 1 * 1 mg
Catalog # ACRBGA1-H4113-1MG
Supplier:  ACROBIOSYSTEMS
Human Galectin-1, Tag Free (GA1-H4113) is expressed from E.coli cells. It contains AA Met 1 - Asp 135 (Accession # P09382-1). 1 * 1 mg
Catalog # ACRBGA1-H4113-1MG
Supplier:  ACROBIOSYSTEMS