ContactUsLinkComponent
Cynomolgus FAP, His Tag (FAP-C52H3) is expressed from human 293 cells (HEK293). It contains AA Leu 26 - Asp 760 (Accession # XP_0055 73377.1). 1 * 100 µG
Catalog # ACRBFAP-C52H3100UG
Supplier:  ACRO Biosystems
Cynomolgus FAP, His Tag (FAP-C52H3) is expressed from human 293 cells (HEK293). It contains AA Leu 26 - Asp 760 (Accession # XP_0055 73377.1). 1 * 100 µG
Catalog # ACRBFAP-C52H3100UG
Supplier:  ACRO Biosystems
Frequently Bought Together