ContactUsLinkComponent
Human TACI (R119A, R122A), Fc Tag (TAI-H5253) is expressed from human 293 cells (HEK293). It contains AA Ser 2 - Thr 166 (Accession # O14836-1 (R119A,R122A)). 1 * 1 mg
Catalog # ACRBTAI-H5253-1MG
Supplier:  ACRO Biosystems
Human TACI (R119A, R122A), Fc Tag (TAI-H5253) is expressed from human 293 cells (HEK293). It contains AA Ser 2 - Thr 166 (Accession # O14836-1 (R119A,R122A)). 1 * 1 mg
Catalog # ACRBTAI-H5253-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together