ContactUsLinkComponent
Cynomolgus 4-1BB Ligand, Fc Tag (41L-C5254) is expressed from human 293 cells (HEK293). It contains AA Leu 67 - Glu 251 (Accession # XP_015296398.1) trimer design. 1 * 100 µG
  ACRB41L-C5254100UG
Cynomolgus 4-1BB Ligand, Fc Tag (41L-C5254) is expressed from human 293 cells (HEK293). It contains AA Leu 67 - Glu 251 (Accession # XP_015296398.1) trimer design. 1 * 100 µG
  ACRB41L-C5254100UG
Frequently Bought Together