ContactUsLinkComponent
Human LAP (TGF-beta 1), Tag Free (LAP-H5213) is expressed from human 293 cells (HEK293). It contains AA Leu 30 - Arg 278 (C33S) (Acc ession # P01137-1 (C33S)). 1 * 1 mg
Catalog # ACRBLAP-H5213-1MG
Supplier:  ACRO Biosystems
Human LAP (TGF-beta 1), Tag Free (LAP-H5213) is expressed from human 293 cells (HEK293). It contains AA Leu 30 - Arg 278 (C33S) (Acc ession # P01137-1 (C33S)). 1 * 1 mg
Catalog # ACRBLAP-H5213-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together