ContactUsLinkComponent
Rhesus macaque DLL3, His Tag (DL3-R52H4) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Arg 490 (Accession # F6 TFV4-1). 1 * 100 µG
Catalog # ACRBDL3-R52H4100UG
Supplier:  ACRO Biosystems
Rhesus macaque DLL3, His Tag (DL3-R52H4) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Arg 490 (Accession # F6 TFV4-1). 1 * 100 µG
Catalog # ACRBDL3-R52H4100UG
Supplier:  ACRO Biosystems
Frequently Bought Together