ContactUsLinkComponent
Mouse CD79B, His Tag (CDB-M52H3) is expressed from human 293 cells (HEK293). It contains AA Val 26 - Asp 158 (Accession # P15530-1). 1 * 100 µG
Catalog # ACRBCDB-M52H3100UG
Supplier:  ACRO Biosystems
Mouse CD79B, His Tag (CDB-M52H3) is expressed from human 293 cells (HEK293). It contains AA Val 26 - Asp 158 (Accession # P15530-1). 1 * 100 µG
Catalog # ACRBCDB-M52H3100UG
Supplier:  ACRO Biosystems
Frequently Bought Together