ContactUsLinkComponent
Biotinylated Human CD79B, His,Avitag (CDB-H82E3) is expressed from human 293 cells (HEK293). It contains AA Ala 29 - Asp 159 (Access ion # P40259-1). 1 * 25 µG
  ACRBCDB-H82E3-25UG
Biotinylated Human CD79B, His,Avitag (CDB-H82E3) is expressed from human 293 cells (HEK293). It contains AA Ala 29 - Asp 159 (Access ion # P40259-1). 1 * 25 µG
  ACRBCDB-H82E3-25UG
Frequently Bought Together