ContactUsLinkComponent
Biotinylated Human CD79B, His,Avitag (CDB-H82E3) is expressed from human 293 cells (HEK293). It contains AA Ala 29 - Asp 159 (Access ion # P40259-1). 1 * 200 µG
Catalog # ACRBCDB-H82E3200UG
Supplier:  ACRO Biosystems
Biotinylated Human CD79B, His,Avitag (CDB-H82E3) is expressed from human 293 cells (HEK293). It contains AA Ala 29 - Asp 159 (Access ion # P40259-1). 1 * 200 µG
Catalog # ACRBCDB-H82E3200UG
Supplier:  ACRO Biosystems
Frequently Bought Together