ContactUsLinkComponent
Biotinylated Human CD2, His,Avitag (CD2-H82E6) is expressed from human 293 cells (HEK293). It contains AA Lys 25 - Asp 209 (Accessio n # AAH33583). 1 * 25 µG
Catalog # ACRBCD2-H82E6-25UG
Supplier:  ACRO Biosystems
Biotinylated Human CD2, His,Avitag (CD2-H82E6) is expressed from human 293 cells (HEK293). It contains AA Lys 25 - Asp 209 (Accessio n # AAH33583). 1 * 25 µG
Catalog # ACRBCD2-H82E6-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together