ContactUsLinkComponent
Biotinylated Human CD72, His,Avitag (CD2-H82E3) is expressed from human 293 cells (HEK293). It contains AA Arg 117 - Asp 359 (Access ion # P21854-1). 1 * 25 µG
Catalog # ACRBCD2-H82E3-25UG
Supplier:  ACRO Biosystems
Biotinylated Human CD72, His,Avitag (CD2-H82E3) is expressed from human 293 cells (HEK293). It contains AA Arg 117 - Asp 359 (Access ion # P21854-1). 1 * 25 µG
Catalog # ACRBCD2-H82E3-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together