ContactUsLinkComponent
Human CD72, His Tag (CD2-H52H3) is expressed from human 293 cells (HEK293). It contains AA Arg 117 - Asp 359 (Accession # P21854-1). 1 * 100 µG
Catalog # ACRBCD2-H52H3100UG
Supplier:  ACRO Biosystems
Human CD72, His Tag (CD2-H52H3) is expressed from human 293 cells (HEK293). It contains AA Arg 117 - Asp 359 (Accession # P21854-1). 1 * 100 µG
Catalog # ACRBCD2-H52H3100UG
Supplier:  ACRO Biosystems
Frequently Bought Together