ContactUsLinkComponent
Human uPAR, Fc Tag (UPR-H5254) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Arg 303 (Accession # Q03405-1). 1 * 1 mg
Catalog # ACRBUPR-H5254-1MG
Supplier:  ACRO Biosystems
Human uPAR, Fc Tag (UPR-H5254) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Arg 303 (Accession # Q03405-1). 1 * 1 mg
Catalog # ACRBUPR-H5254-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together