Orders LinkContactUsLinkComponent
Accessories for multi-use data loggers, tempmate.®-M2
tempmate.®-M2 external T/rH-sensor –40...+90 °C HQ digital temperature/humidity sensor 1,2 m length, 0 - 100% RH
Catalog # IMECM2S-200
Supplier:   tempmate
Accessories for multi-use data loggers, tempmate.®-M2
Catalog # IMECM2S-200
Supplier:  tempmate
Accessories for multi-use data loggers, tempmate.®-M2
Accessories for multi-use data loggers, tempmate.®-M2
tempmate.®-M2 external T/rH-sensor –40...+90 °C HQ digital temperature/humidity sensor 1,2 m length, 0 - 100% RH
Price
26,20 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    tempmate.®-M2 external T/rH-sensor –40...+90 °C HQ digital temperature/humidity sensor 1,2 m length, 0 - 100% RH
  • Seq:
    02