ContactUsLinkComponent
Biotinylated SARS-CoV-2 Spike RBD (K417N, L452R, T478K), His,Avitag (SPD-C82Eg) is expressed from human 293 cells (HEK293). It conta ins AA Arg 319 - Lys 537 (Accession QHD43416.1 (K417N, L452R, T478K)). The mutations K417N, L452R, T478K are identif 1 * 200 µG
Catalog # ACRBSPD-C82EG200UG
Supplier:  ACROBIOSYSTEMS
Biotinylated SARS-CoV-2 Spike RBD (K417N, L452R, T478K), His,Avitag (SPD-C82Eg) is expressed from human 293 cells (HEK293). It conta ins AA Arg 319 - Lys 537 (Accession QHD43416.1 (K417N, L452R, T478K)). The mutations K417N, L452R, T478K are identif 1 * 200 µG
Catalog # ACRBSPD-C82EG200UG
Supplier:  ACROBIOSYSTEMS