ContactUsLinkComponent
SARS-CoV-2 Spike RBD (L452R, T478K), His Tag (SPD-C52Hh) is expressed from human 293 cells (HEK293). It contains AA Arg 319 - Lys 53 7 (Accession QHD43416.1 (L452R, T478K)). The mutations (L452R, T478K) were identified in the SARS-CoV-2 variants whi 1 * 100 µG
Catalog # ACRBSPD-C52HH100UG
Supplier:  ACROBIOSYSTEMS
SARS-CoV-2 Spike RBD (L452R, T478K), His Tag (SPD-C52Hh) is expressed from human 293 cells (HEK293). It contains AA Arg 319 - Lys 53 7 (Accession QHD43416.1 (L452R, T478K)). The mutations (L452R, T478K) were identified in the SARS-CoV-2 variants whi 1 * 100 µG
Catalog # ACRBSPD-C52HH100UG
Supplier:  ACROBIOSYSTEMS