ContactUsLinkComponent
Vial & Cap Kit. 100 EA 2ml, Screw Top, Hydrophobic, Amber Autosampler Vials and matching Black Caps with Clear Pre-Slit Silicone Rub ber / PTFE Septa. RSA-Pro X Brand. 1 PK. 1 * 100 items
Catalog # MICS9509S-3DAP-PX
Supplier:  MICROSOLV
Vial & Cap Kit. 100 EA 2ml, Screw Top, Hydrophobic, Amber Autosampler Vials and matching Black Caps with Clear Pre-Slit Silicone Rub ber / PTFE Septa. RSA-Pro X Brand. 1 PK. 1 * 100 items
Catalog # MICS9509S-3DAP-PX
Supplier:  MICROSOLV