ContactUsLinkComponent
Sodium Standard for Ion Chromatography (IC) 1 mg/mL (1000 ppm) in 0.005% Nitric Acid (HNO2) 1 * 500 mL
Catalog # REGEICCB04
Supplier:  REAGECON
Sodium Standard for Ion Chromatography (IC) 1 mg/mL (1000 ppm) in 0.005% Nitric Acid (HNO2) 1 * 500 mL
Catalog # REGEICCB04
Supplier:  REAGECON
Frequently Bought Together