Blower transition adapters
Blower transition adapter, 254,0 mm nominal diameter, 1,4 kg
Catalog # LBCP4861700
Supplier: LABCONCO
Wechseln zu:
Product Details & Documents
Epoxy-coated steel blower transition adapters fit all Labconco coated steel blowers.
Adapter allows you to connect thermoplastic duct to the exhaust side of a blower to create an exhaust stack. A nominal size PVC duct fits inside the opening.
Spezifikationen:
- Description:Blower transition adapter, 254,0 mm nominal diameter
- Weight:1,4 kg



