ContactUsLinkComponent
SARS-COV-2 SPIKE TRIMER (T19R, G142D, EF156-157DEL, R158G, L452R, T478K, D614G, P681R, D950N), HIS TAG (MALS VERIFIED) 1 * 500 µG
Catalog # ACRBSPN-C52HE500UG
Supplier:  ACRO Biosystems
SARS-COV-2 SPIKE TRIMER (T19R, G142D, EF156-157DEL, R158G, L452R, T478K, D614G, P681R, D950N), HIS TAG (MALS VERIFIED) 1 * 500 µG
Catalog # ACRBSPN-C52HE500UG
Supplier:  ACRO Biosystems
Frequently Bought Together