Fluorescamine
Bottle, 1 g
APOSBIF4008-1G
Reacts with primary amines to form stable, highlyfluorescent products.
: Fluram, 4-Phenyl-3H,3'H-spiro[furan-2,1'-isobenzofuran]-3,3'-dione, 4-Phenylspiro[furan-2(3H),1'-phthalan]-3,3'-dione
- :38183-12-9
- :C₁₇H₁₀O₄
- :MFCD00005928
- :278.26 g/mol
- :Ambient
- Pack type:Bottle
- Pk:1 g