Fluorescamine
Bottle, 250 mg
Catalog # APOSBIF4008-250MG
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Reacts with primary amines to form stable, highlyfluorescent products.
Synonyms: Fluram, 4-Phenyl-3H,3'H-spiro[furan-2,1'-isobenzofuran]-3,3'-dione, 4-Phenylspiro[furan-2(3H),1'-phthalan]-3,3'-dione
- CAS Number:38183-12-9
- Formula:C₁₇H₁₀O₄
- MDL Number:MFCD00005928
- Molecular Weight:278.26 g/mol
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Bottle
- Pk:250 mg



