Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Cleanroom spiral college notebook DIN A5 squared, 72 gr/m, 50 sheets color: light blue RR-class: 100 (ISO 5) 1 * 5 ST
Catalog # MSPPRPSPIRAL-A5BK
Supplier:
MARKET SOURCE PART PROCESS
Expand image
Cleanroom spiral college notebook DIN A5 squared, 72 gr/m, 50 sheets color: light blue RR-class: 100 (ISO 5) 1 * 5 ST
Catalog # MSPPRPSPIRAL-A5BK
Supplier:
MARKET SOURCE PART PROCESS