ContactUsLinkComponent
4 945 202 Centrifuge tubes, 25 ml, with round bottom, ungraduated, heavy wall, with screw cap, o.d. 24 mm, length 98 mm, nominal volume 20 ml 1 * 1 items
  WITG4.945.202
4 945 202 Centrifuge tubes, 25 ml, with round bottom, ungraduated, heavy wall, with screw cap, o.d. 24 mm, length 98 mm, nominal volume 20 ml 1 * 1 items
  WITG4.945.202
Frequently Bought Together