Human Recombinant FGF2 (from E. coli)
Catalog # ABCAAB61845-100
Marketsource
Supplier: Abcam
Human Recombinant FGF2 (from E. coli)
Catalog # ABCAAB61845-100
Supplier: Abcam
Marketsource
CAS Number:
Jump to:
Product Details & Documents
Recombinant human FGF2 protein is a Human Full Length protein, expressed in Escherichia coli, with >95% purity and suitable for SDS-PAGE, FuncS.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Specifications
Specifications
- Form:Lyophilised
- Protein/peptide name:FGF2
- Protein synonyms:FGFB|Fibroblast growth factor 2|FGF-2|Basic fibroblast growth factor|Heparin-binding growth factor 2|bFGF|HBGF-2
- Purity:>95%
- Source:E. coli
- Biological activity:Standard
- Shipping temperature:20
- Cat. No.:ABCAAB61845-100
- Protein/peptide type:Recombinant
- Storage conditions:Store at −20 °C
- Formulation:Constituents: 0,164% Sodium phosphate
- Conjugation:Unconjugated
- Tested applications:FuncS, SDS-PAGE
- Species:Human
- Animal-free:N



