Order Entry
Germany
Orders LinkContactUsLinkComponent
Multiple affinity removal column human 14

Multiple affinity removal column human 14

Multiple affinity removal column, human-14, 4,60 mm, 100 mm
Catalog # AGLS5188-6558
Supplier:   AGILENT
Multiple affinity removal column human 14
Catalog # AGLS5188-6558
Supplier:  AGILENT
Multiple affinity removal column human 14
Multiple affinity removal column human 14

Multiple affinity removal column, human-14, 4,60 mm, 100 mm

Price
8 137,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

The Human 14 Multiple Affinity Removal Column is designed to chromatographically remove fourteen interfering high-abundant proteins from human plasma samples.

  • Made of PEEK (polyetheretherketone)
  • High-capacity serum load
  • Affinity resin packed

Removal of these abundant proteins improves the subsequent LC/MS and electrophoretic analysis of the serum sample by effectively expanding the dynamic range of the analysis.

Specifications
  • Ø int.:
    4,60 mm
  • Description:
    Multiple affinity removal column, human-14
  • Length:
    100 mm
  • Seq:
    02