Multiple affinity removal column human 14
Multiple affinity removal column, human-14, 4,60 mm, 50 mm
Catalog # AGLS5188-6557
Supplier: AGILENT
Jump to:
Product Details & Documents
The Human 14 Multiple Affinity Removal Column is designed to chromatographically remove fourteen interfering high-abundant proteins from human plasma samples.
- Made of PEEK (polyetheretherketone)
- High-capacity serum load
- Affinity resin packed
Removal of these abundant proteins improves the subsequent LC/MS and electrophoretic analysis of the serum sample by effectively expanding the dynamic range of the analysis.
Specifications
- Ø int.:4,60 mm
- Description:Multiple affinity removal column, human-14
- Length:50 mm
- Seq:01
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



