Order Entry
Germany
Orders LinkContactUsLinkComponent
Multiple affinity removal starter reagent kits

Multiple affinity removal starter reagent kits

Spin Concentrators, 4 ml, to concentrate proteins via low-speed centrifugation through 5K MWCo filters into a collection tube, 25
Catalog # AGLS5185-5991
Supplier:   AGILENT
Multiple affinity removal starter reagent kits
Catalog # AGLS5185-5991
Supplier:  AGILENT
CAS Number:  
Multiple affinity removal starter reagent kits
Multiple affinity removal starter reagent kits

Spin Concentrators, 4 ml, to concentrate proteins via low-speed centrifugation through 5K MWCo filters into a collection tube, 25

CAS Number:  
Price
248,00 €
Log in for availability
1 * 25 ST
Product Details & Documents

Starter reagent kit for multiple affinity removal LC columns.

Delivery information: Includes: 2 bottles buffer A (1 L), 1 bottle buffer B (1 L), 2 packs of 25 spin filters, 1 pack of 25 spin concentrator.

Specifications

Specifications

  • Pk:
    25
  • Description:
    Spin Concentrators, 4 ml
  • Details:
    to concentrate proteins via low-speed centrifugation through 5K MWCo filters into a collection tube

Specifications