Orders LinkContactUsLinkComponent
Multiple affinity removal starter reagent kits
Multiple affinity removal starter reagent kits
Multiple affinity removal buffer B, 1 L, For eluting, 1
Catalog # AGLS5185-5988
Supplier:   AGILENT
Multiple affinity removal starter reagent kits
Catalog # AGLS5185-5988
Supplier:  AGILENT
CAS Number:  
Multiple affinity removal starter reagent kits
Multiple affinity removal starter reagent kits
Multiple affinity removal buffer B, 1 L, For eluting, 1
CAS Number:  
Price
168,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Starter reagent kit for multiple affinity removal LC columns.
Delivery information: Includes: 2 bottles buffer A (1 L), 1 bottle buffer B (1 L), 2 packs of 25 spin filters, 1 pack of 25 spin concentrator.
Specifications
Specifications
  • Pk:
    1
  • Description:
    Multiple affinity removal buffer B, 1 L
  • Details:
    For eluting
Specifications