Multiple affinity removal starter reagent kits
Multiple affinity removal reagent kit, -, 1 kt
Catalog # AGLS5185-5986
Supplier: AGILENT
Jump to:
Product Details & Documents
Starter reagent kit for multiple affinity removal LC columns.
Delivery information: Includes: 2 bottles buffer A (1 L), 1 bottle buffer B (1 L), 2 packs of 25 spin filters, 1 pack of 25 spin concentrator.
Specifications
Specifications
- VE:1 kt
- Bezeichnung:Multiple affinity removal reagent kit
- Details:-