Wire baskets, round, with handle
Wire basket, round, with handle, 100×80 mm
Catalog # 216-2376
Supplier: RSG
Jump to:
Product Details & Documents
Made from stainless steel 1.4301.
- Rust-proof
- Very good temperature resistance
- Good chemical resistance
- With reinforced edges
Specifications
- Ø×H:100×80 mm
- Description:Wire basket, round, with handle
- Seq:01