Order Entry
Germany
Orders LinkContactUsLinkComponent
Double micro clamps

Double micro clamps

Variable clamp press, 5 - 15 g, Stainless steel, 7 mm, Straight, 3,5×1 mm
  FSCI18040-11
Marketsource
Double micro clamps
  FSCI18040-11
Marketsource
Double micro clamps
Double micro clamps

Variable clamp press, 5 - 15 g, Stainless steel, 7 mm, Straight, 3,5×1 mm

 
495,00 €
   
1 * 1 ST
 
 
  • Description:
    Variable clamp press, 5 - 15 g
  • Length:
    7 mm
  • Material:
    Stainless steel
  • Seq:
    01
  • Tip size:
    3,5×1 mm
  • Tip shape:
    Straight