Order Entry
Germany
Orders LinkContactUsLinkComponent
Fine forceps, self-closing

Fine forceps, self-closing

Stainless steel, 115 mm, Straight, 0,8×0,04 mm
Catalog # FSCI11480-11
Marketsource
Supplier:   FINE SCIENCE TOOLS
Fine forceps, self-closing
Catalog # FSCI11480-11
Supplier:  FINE SCIENCE TOOLS
Marketsource
Fine forceps, self-closing
Fine forceps, self-closing

Stainless steel, 115 mm, Straight, 0,8×0,04 mm

Price
93,20 €
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Length:
    115 mm
  • Material:
    Stainless steel
  • Seq:
    02
  • Tip size:
    0,8×0,04 mm
  • Tip shape:
    Straight