Order Entry
Germany
Orders LinkContactUsLinkComponent
Grounding cables, b.safe

Grounding cables, b.safe

Grounding cable with clip and clip for tube Ø 32 mm, 3,0 m
Catalog # 590-0342
Supplier:   Bohlender
Grounding cables, b.safe
Catalog # 590-0342
Supplier:  Bohlender
Grounding cables, b.safe
Grounding cables, b.safe

Grounding cable with clip and clip for tube Ø 32 mm, 3,0 m

Price
32,70 €
Log in for availability
1 * 1 ST
Product Details & Documents

For earth connection of vessels and components made of static dissipative materials.

  • Choose the suitable connection to earth that we offer in our range of grounding cables with different connectors
  • Customised versions can be offered on request
  • Suitable grounding accessories for b.safe products

The scope of delivery of the ring cable lug includes screw, nut and washer.

Specifications
  • Description:
    Grounding cable with clip and clip for tube Ø 32 mm
  • Length:
    3,0 m
  • Seq:
    08
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.