Capillary cutter, b.safe
Capillary cutter with guide bores for tubing up to Ext.Ø 8 mm
Catalog # 590-0334
Supplier: Bohlender
Jump to:
Product Details & Documents
Ideal for cutting capillaries and elastic tubes without reinforcement.
- Capillary cutter for up to tubing Ext.Ø 8 mm with guide bores
- High qualitiy capillary cutter
- Replacement blades available
The guide bores help to get a clean cut of the capillaries.
Specifications
- Description:Capillary cutter with guide bores for tubing up to Ext.Ø 8 mm
- Seq:01