Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Phosphomolybdic acid hydrate ACS

Bottle
Catalog # APOSIN2835-100G
Oxidizing
Health
Corrosive
Supplier:   Apollo Scientific
Phosphomolybdic acid hydrate ACS
Catalog # APOSIN2835-100G
Supplier:  Apollo Scientific
CAS Number:  51429-74-4
Phosphomolybdic acid hydrate ACS

Bottle

CAS Number:  51429-74-4
Price
239,00 €
Log in for availability
1 * 100 g
Product Details & Documents

Usage: Inorganic salt or catalyst.

Synonyms:  Molybdophosphoric acid hydrate, 12-Molybdophosphoric acid hydrate, Phosphomolybdic acid hydrate
  • CAS Number:51429-74-4
  • Formula:12MoO₃·H₃PO₄·H₂O
  • MDL Number:MFCD00011341
  • Molecular Weight:1843,26886 g/mol
  • EINECS:234-713-5
  • UN:3085
Specifications

Specifications

  • Pack type:
    Bottle

Specifications