Order Entry
Germany
Orders LinkContactUsLinkComponent
4-Pentylcyclohexanone 98%

4-Pentylcyclohexanone 98%

Bottle, 5 g
Catalog # APOSOR914341-5G
Supplier:   Apollo Scientific
4-Pentylcyclohexanone 98%
Catalog # APOSOR914341-5G
Supplier:  Apollo Scientific
CAS Number:  61203-83-6
4-Pentylcyclohexanone 98%

Bottle, 5 g

CAS Number:  61203-83-6
Price
29,00 €
Log in for availability
1 * 5 g
Product Details & Documents
  • CAS Number:61203-83-6
  • Formula:C₁₁H₂₀O
  • MDL Number:MFCD00059581
  • Molecular Weight:168.28 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.