Order Entry
Germany
Orders LinkContactUsLinkComponent
2-(2-Bromophenyl)ethanol 98%

2-(2-Bromophenyl)ethanol 98%

Bottle, 100 g
Catalog # APOSOR480688-100G
Irritant
Supplier:   Apollo Scientific
2-(2-Bromophenyl)ethanol 98%
Catalog # APOSOR480688-100G
Supplier:  Apollo Scientific
CAS Number:  1074-16-4
2-(2-Bromophenyl)ethanol 98%

Bottle, 100 g

CAS Number:  1074-16-4
Price
180,00 €
Log in for availability
1 * 100 g
Product Details & Documents
  • CAS Number:1074-16-4
  • Formula:C₈H₉BrO
  • MDL Number:MFCD00093566
  • Molecular Weight:201.06 g/mol
  • Boiling Pt:97 °C/0.675mmHg
  • Density:1.483 g/cm³ (25 °C)
  • Flash Pt:>113 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.