Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Acetyl iodide 95%

Bottle, 10 g
Catalog # APOSOR938283-10G
Supplier:   Apollo Scientific
Acetyl iodide 95%
Catalog # APOSOR938283-10G
Supplier:  Apollo Scientific
CAS Number:  507-02-8
Acetyl iodide 95%

Bottle, 10 g

CAS Number:  507-02-8
Price
963,00 €
Log in for availability
1 * 10 g
Product Details & Documents
  • CAS Number:507-02-8
  • Formula:C₂H₃IO
  • MDL Number:MFCD00039399
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    10 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.