Order Entry
Germany
Orders LinkContactUsLinkComponent
5'-Chloro-2'-hydroxypropiophenone

5'-Chloro-2'-hydroxypropiophenone

Bottle, 5 g
Catalog # APOSOR22404-5G
Irritant
Supplier:   Apollo Scientific
5'-Chloro-2'-hydroxypropiophenone
Catalog # APOSOR22404-5G
Supplier:  Apollo Scientific
CAS Number:  2892-16-2
5'-Chloro-2'-hydroxypropiophenone

Bottle, 5 g

CAS Number:  2892-16-2
Price
128,00 €
Log in for availability
1 * 5 g
Product Details & Documents
  • CAS Number:2892-16-2
  • Formula:C₉H₉ClO₂
  • MDL Number:MFCD00218579
  • Melting Pt:35 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.