Order Entry
Germany
Orders LinkContactUsLinkComponent
2-(Methylthio)nicotinoyl chloride

2-(Methylthio)nicotinoyl chloride

Bottle, 5 g
Catalog # APOSOR21399-5G
Corrosive
Supplier:   Apollo Scientific
2-(Methylthio)nicotinoyl chloride
Catalog # APOSOR21399-5G
Supplier:  Apollo Scientific
CAS Number:  97936-43-1
2-(Methylthio)nicotinoyl chloride

Bottle, 5 g

CAS Number:  97936-43-1
Price
484,00 €
Log in for availability
1 * 5 g
Product Details & Documents
  • CAS Number:97936-43-1
  • Formula:C₇H₆ClNOS
  • MDL Number:MFCD00051678
  • Molecular Weight:187.65 g/mol
  • Melting Pt:92…96 °C
  • Storage Temperature:Ambient
  • UN:3261
  • ADR:8,II
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.