Order Entry
Germany
Orders LinkContactUsLinkComponent
Thioisonicotinamide 98%

Thioisonicotinamide 98%

Bottle, 100 g
Catalog # APOSOR903374-100G
Supplier:   Apollo Scientific
Thioisonicotinamide 98%
Catalog # APOSOR903374-100G
Supplier:  Apollo Scientific
CAS Number:  2196-13-6
Thioisonicotinamide 98%

Bottle, 100 g

CAS Number:  2196-13-6
Price
99,90 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  Pyridine-4-carbothioamide
  • CAS Number:2196-13-6
  • Formula:C₆H₆N₂S
  • MDL Number:MFCD00006437
  • Molecular Weight:138.19 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.