Order Entry
Germany
Orders LinkContactUsLinkComponent
 

6-Hydroxyindole 95%

Bottle, 250 mg
Catalog # APOSOR7612-250MG
Corrosive
Irritant
Environment
Supplier:   Apollo Scientific
6-Hydroxyindole 95%
Catalog # APOSOR7612-250MG
Supplier:  Apollo Scientific
CAS Number:  2380-86-1
6-Hydroxyindole 95%

Bottle, 250 mg

CAS Number:  2380-86-1
Price
21,80 €
Log in for availability
1 * 250 mg
Product Details & Documents
Synonyms:  Indol-6-ol, 6-Indolol, IMEXINE OBA, 6-Hydroxy-1H-indole, 1H-Indol-6-ol
  • CAS Number:2380-86-1
  • Formula:C₈H₇NO
  • MDL Number:MFCD00152101
  • Molecular Weight:133.15 g/mol
  • Melting Pt:125…128 °C
  • Storage Temperature:Refrigerator
  • EINECS:417-020-4
  • UN:3077
  • ADR:9+EHS,III
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg

Specifications