Order Entry
Germany
Orders LinkContactUsLinkComponent
2-Fluoro-5-nitroanisole

2-Fluoro-5-nitroanisole

Bottle, 25 g
Catalog # APOSPC4443-25G
Irritant
Supplier:   Apollo Scientific
2-Fluoro-5-nitroanisole
Catalog # APOSPC4443-25G
Supplier:  Apollo Scientific
CAS Number:  454-16-0
2-Fluoro-5-nitroanisole

Bottle, 25 g

CAS Number:  454-16-0
Price
88,30 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  1-Fluoro-2-methoxy-4-nitrobenzene, 4-Fluoro-3-methoxynitrobenzene
  • CAS Number:454-16-0
  • Formula:C₇H₆FNO₃
  • MDL Number:MFCD07782081
  • Molecular Weight:171.13 g/mol
  • Melting Pt:66…71 °C
  • Flash Pt:>110 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.