Order Entry
Germany
Orders LinkContactUsLinkComponent
1-Hydroxypiperidine 95%

1-Hydroxypiperidine 95%

Bottle, 5 g
Catalog # APOSOR961933-5G
Supplier:   Apollo Scientific
1-Hydroxypiperidine 95%
Catalog # APOSOR961933-5G
Supplier:  Apollo Scientific
CAS Number:  4801-58-5
1-Hydroxypiperidine 95%

Bottle, 5 g

CAS Number:  4801-58-5
Price
478,00 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  Piperidin-1-ol
  • CAS Number:4801-58-5
  • Formula:C₅H₁₁NO
  • MDL Number:MFCD00038048
  • Molecular Weight:101.147 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.