Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
PAB solution for sanitization of ProSep and Eshmuno Protein A affinity chromatography media 1 * 1 L
4.80949.1000
:
Merck
PAB solution for sanitization of ProSep and Eshmuno Protein A affinity chromatography media 1 * 1 L
4.80949.1000
:
Merck